Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOL8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103219 Supplier Invitrogen™ Supplier No. PA5103219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NOL8.

NOL8 binds Ras-related GTP-binding proteins and plays a role in cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NOL8
Gene Accession No. Q3UHX0, Q76FK4
Gene Alias 4921532D18Rik; 5730412B09Rik; bA62C3.3; bA62C3.4; C9orf34; NOL8; NOP132; Nucleolar protein 8; nucleolar protein Nop132
Gene Symbols NOL8
Host Species Rabbit
Immunogen A synthesized peptide derived from human NOL8(Accession Q76FK4), corresponding to amino acid residues G1134-K1167.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55035, 70930
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.