Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOL8 Polyclonal Antibody

Catalog No. PIPA535937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535937 Supplier Invitrogen™ Supplier No. PA535937
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HeLa nuclear. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NOL8 binds Ras-related GTP-binding proteins and plays a role in cell growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL8
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NOL8
Gene Accession No. Q76FK4
Gene Alias 4921532D18Rik; 5730412B09Rik; bA62C3.3; bA62C3.4; C9orf34; NOL8; NOP132; Nucleolar protein 8; nucleolar protein Nop132
Gene Symbols NOL8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human NOL8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.