Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOL8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551027
Description
NOL8 Polyclonal Antibody detects endogenous levels of NOL8 protein.
NOL8 binds Ras-related GTP-binding proteins and plays a role in cell growth.
Specifications
NOL8 | |
Polyclonal | |
Unconjugated | |
NOL8 | |
4921532D18Rik; 5730412B09Rik; bA62C3.3; bA62C3.4; C9orf34; NOL8; NOP132; Nucleolar protein 8; nucleolar protein Nop132 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55035 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q76FK4 | |
NOL8 | |
Synthesized peptide derived from the C-terminal region of human Nop132. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction