Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103988 Supplier Invitrogen™ Supplier No. PA5103988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NOL9.

NOL9 is a polynucleotide 5'-kinase involved in rRNA processing. The kinase activity is required for the processing of the 32S precursor into 5.8S and 28S rRNAs, more specifically for the generation of the major 5.8S(S) form. In vitro, it has both DNA and RNA 5'-kinase activities and probably binds RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NOL9
Gene Accession No. Q3TZX8, Q5SY16
Gene Alias 4632412I24Rik; 6030462G04Rik; AI449622; AW490720; Grc3; NET6; NOL9; Nucleolar protein 9; polynucleotide 5'-hydroxyl-kinase NOL9; polynucleotide 5'-kinase
Gene Symbols NOL9
Host Species Rabbit
Immunogen A synthesized peptide derived from human NOL9(Accession Q5SY16), corresponding to amino acid residues R317-T367.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313744, 74035, 79707
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.