Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOLA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143962
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143962 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143962 Supplier Invitrogen™ Supplier No. PA5143962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human A375 whole cell, human K562 whole cell, human HL-60 whole cell, human MCF-7 whole cell. IHC: human breast cancer tissue, human gastric carcinoma tissue, human liver cancer tissue, human ovarian adenoma tissue, human pancreatic cancer tissue, human prostate cancer tissue, human glioma tissue, human lymphoma tissue. ICC/IF: MCF-7 cell. Flow: K562 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Required for ribosome biogenesis and telomere maintenance. NOLA1 is part of the H/ACA small nucleolar ribonucleoprotein (H/ACA snoRNP) complex, which catalyzes pseudouridylation of rRNA. This involves the isomerization of uridine such that the ribose is subsequently attached to C5, instead of the normal N1. Each rRNA can contain up to 100 pseudouridine ('psi') residues, which may serve to stabilize the conformation of rRNAs. NOLA1 may also be required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOLA1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene GAR1
Gene Accession No. Q9NY12
Gene Alias AA409823; AI326794; C430047J18Rik; Gar1; GAR1 homolog, ribonucleoprotein; GAR1 ribonucleoprotein; GAR1 ribonucleoprotein homolog; GAR1 ribonucleoprotein homolog (yeast); H/ACA ribonucleoprotein complex subunit 1; H/ACA small nucleolar RNPs; Nola1; Nucleolar protein family A member 1; nucleolar protein family A, member 1 (H/ACA small nucleolar RNPs); snoRNP protein GAR1
Gene Symbols GAR1
Host Species Rabbit
Immunogen E.coli-derived human NOLA1/GAR1 recombinant protein (Position: F58-K165).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54433
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.