Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOLC1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP122981AF488
Description
NOLC1 Polyclonal antibody specifically detects NOLC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NOLC1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
140 kDa nucleolar phosphoprotein, HCV NS5A trans-regulated protein 13, HCV NS5A-transactivated protein 13, Hepatitis C virus NS5A-transactivated protein 13, KIAA0035NS5ATP13, NOPP130, NOPP140, Nucleolar 130 kDa protein, nucleolar and coiled-body phosphoprotein 1, nucleolar and coiled-body phosphprotein 1, Nucleolar phosphoprotein p130, nucleolar protein p130, P130 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human nucleolar and coiled-body phosphoprotein 1 using the numbering given in entry NP_004732.2 (GeneID 9221). | |
0.1 mL | |
Cell Cycle and Replication | |
9221 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction