Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOLC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535938
Description
Recommended positive controls: H1299, HCT116, K562, HL-60. Predicted reactivity: Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Related to nucleologenesis, may play a role in the maintenance of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It has intrinsic GTPase and ATPase activities. May play an important role in transcription catalyzed by RNA polymerase I.
Specifications
| NOLC1 | |
| Polyclonal | |
| Unconjugated | |
| NOLC1 | |
| 140 kDa nucleolar phosphoprotein; HCV NS5A trans-regulated protein 13; HCV NS5A-transactivated protein 13; Hepatitis C virus NS5A-transactivated protein 13; KIAA0035; NOLC1; nolc1.L; nopp130; nopp140; NS5ATP13; Nucleolar 130 kDa protein; nucleolar and coiled-body phosphoprotein 1; nucleolar and coiled-body phosphoprotein 1 L homeolog; nucleolar and coiled-body phosphprotein 1; nucleolar phosphoprotein; nucleolar phosphoprotein 140; nucleolar phosphoprotein p130; nucleolar protein p130; P130; XELAEV_18034881mg; xNopp180 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9221 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q14978 | |
| NOLC1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human NOLC1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction