Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOLC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596472 Supplier Invitrogen™ Supplier No. PA596472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKPAAAPKQP VGGGQKLLTR KADSSSSEEE SSSSEEEKTK KMVATTKPKA TAKAALSLPA KQAPQGSRDS SSDSDSSSSE EEEEKTSKSA VKKKPQKVAG GAAPSKPASA KKGKAESSNS SSSDDSSEEE EEKLKGKGSP RPQAPKANGT SALTAQNGKA AKNSEEEEEE KKKAAVVVSK SGSLKKRKQN EAAKEAETPQ AKKIKLQTPN TFPKRKKGEK RASSPFRRVR EEEIEVDSRV ADNSFDAKRG AAGDWGERAN QVLKFTKGKS FRHEKTKKKR GSYRGGSISV QVNSIKFDSE; Positive Samples: A-549, HeLa, HepG2, MCF7; Cellular Location: Cytoplasm, Nucleus, nucleolus.

Related to nucleologenesis, may play a role in the maintenance of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It has intrinsic GTPase and ATPase activities. May play an important role in transcription catalyzed by RNA polymerase I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOLC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NOLC1
Gene Accession No. E9Q5C9, P41777, Q14978
Gene Alias 140 kDa nucleolar phosphoprotein; HCV NS5A trans-regulated protein 13; HCV NS5A-transactivated protein 13; Hepatitis C virus NS5A-transactivated protein 13; KIAA0035; NOLC1; nolc1.L; nopp130; nopp140; NS5ATP13; Nucleolar 130 kDa protein; nucleolar and coiled-body phosphoprotein 1; nucleolar and coiled-body phosphoprotein 1 L homeolog; nucleolar and coiled-body phosphprotein 1; nucleolar phosphoprotein; nucleolar phosphoprotein 140; nucleolar phosphoprotein p130; nucleolar protein p130; P130; XELAEV_18034881mg; xNopp180
Gene Symbols NOLC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-699 of human NOLC1 (NP_0047322).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64896, 70769, 9221
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.