Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOLC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | NOLC1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-7477
![]() ![]() |
Proteintech Group Inc
118151AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
NOPP140 (140 kDa nucleolar phosphoprotein) is implicated to play a role in ribosome assembly and the regulation of rRNA transcription. It has been observed to shuttle between the nucleus and cytoplasm and may also be important to nucleolar structural organization.Specifications
NOLC1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
E9Q5C9, P41777, Q14978 | |
64896, 70769, 9221 | |
NOLC1 Fusion Protein Ag2382 | |
Primary | |
-20°C | |
NOLC1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA0035, NOLC1, NOPP130, NOPP140, NS5ATP13, Nucleolar 130 kDa protein, Nucleolar phosphoprotein p130 | |
Nolc1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title