Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOMO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583833
Description
Immunogen sequence: GEKITVTPSS KELLFYPPSM EAVVSGESCP GKLIEIHGKA GLFLEGQIHP ELEGVEIVIS EKGASSPLIT VFTDDKGAYS VGPLHSDLEY TVTS.
This gene encodes a protein originally thought to be related to the collagenase gene family. This gene is one of three highly similar genes in a region of duplication located on the p arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development. Mutations in ABCC6, which is located nearby, rather than mutations in this gene are associated with pseudoxanthoma elasticum.
Specifications
NOMO1 | |
Polyclonal | |
Unconjugated | |
NOMO1 | |
D7Ertd156e; Nodal modulator 1; nodal modulator 3; Nomo; NOMO1; PM5; pM5 protein; pM5 protein 3; pM5 protein, telomeric copy; RGD1305240 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23420 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15155 | |
NOMO1 | |
Recombinant protein corresponding to Human NOMO1. Recombinant protein control fragment (Product #RP-104407). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction