Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOMO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595819
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595819 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595819 Supplier Invitrogen™ Supplier No. PA595819
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YALAGVSFEI KAEDDQPLPG VLLSLSGGLF RSNLLTQDNG ILTFSNLSPG QYYFKPMMKE FRFEPSSQMI EVQEGQNLKI TITGYRTAYS CYGTVSSLNG EPEQGVAMEA VGQNDCSIYG EDTVTDEEGK FRLRGLLPGC VYHVQLKAEG NDHIERALPH HRVIEVGNND IDDVNIIVFR QINQFDLSGN VITSSEYLPT LWVKLYKSEN LDNPIQTVSL GQSLFFHFPP LLRDGENYVV LLDSTLPRSQ YDYILPQVSF TAVGYHKHIT LIFNPTRKLP EQDIAQGSYI ALPLTLLVLL AGYNHDKLIP LLLQLTSRLQ GVGALGQAAS DNSGPEDAKR QAKKQKTRRT; Positive Samples: BxPC3; Cellular Location: Endoplasmic reticulum membrane, Single-pass membrane protein.

This gene encodes a protein originally thought to be related to the collagenase gene family. This gene is one of three highly similar genes in a region of duplication located on the p arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development. Mutations in ABCC6, which is located nearby, rather than mutations in this gene are associated with pseudoxanthoma elasticum (PXE). Two transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOMO2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NOMO2
Gene Accession No. Q5JPE7
Gene Alias nodal modulator 2; Nomo; NOMO2; PM5; pM5 protein 2; pM5 protein, centromeric copy
Gene Symbols NOMO2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 873-1222 of human NOMO2 (NP_0010040601).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283820
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.