Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOMO3 Monoclonal Antibody (5A7)
GREENER_CHOICE

Catalog No. PIMA524540
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524540 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524540 Supplier Invitrogen™ Supplier No. MA524540
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: TVSSLNGEPE QGVAMEAVGQ NDCSIYGEDT VTDEEGKFRL RGLLPGCVYH VQLKAEGNDH IERALPHH.

This gene encodes a protein originally thought to be related to the collagenase gene family. This gene is one of three highly similar genes in a duplicated region on the short arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development. Mutations in ABCC6, which is located nearby, rather than mutations in this gene are associated with pseudoxanthoma elasticum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOMO3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene NOMO3
Gene Accession No. P69849
Gene Alias nodal modulator 3; Nomo; NOMO3; pM5 protein 3; pM5 protein, middle copy
Gene Symbols NOMO3
Host Species Mouse
Immunogen NOMO3 (NP_001004067.1, 966 a.a. ~ 1033 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 408050
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.