Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Non-human Primate GM-CSF ELISA Kit
SDP

Catalog No. NB397904 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit (1 Plate)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397904 1 Kit (1 Plate)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397904 Supplier Novus Biologicals™ Supplier No. NBP311733

ELISA Kit

Specifications

Assay Range 3.16 to 1000 pg/mL
Assay Sensitivity 1 pg/mL
Immunoassay Kit Format ELISA (Standard)
Product Type ELISA Kit
Sample Type Serum, Plasma, Cell Culture Supernatant
Target Species Non-human Primate
For Use With (Application) ELISA
Gene ID (Entrez) 1437
Gene Symbol CSF2
Quantity 1 Kit (1 Plate)
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Target Non-human Primate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.