Learn More
Specifications
Specifications
| Assay Range | 12.21 to 50000 pg/mL |
| Assay Sensitivity | 2.9 pg/mL |
| Immunoassay Kit Format | Colorimetric Detection Kit |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Culture Supernatant, Plasma, Serum, Saliva, Urine |
| Specificity | Cross reactivities for a number of related steroid compounds: DHEA sulfate 30% Androstenedione 0.72% Androsterone 0.29% Pregnenolone 0.28% Testosterone 0.10% Progesterone 0.06% Estradiol 0.04% 20 alpha-hydroxycholesterol <0.02% Androsterone sulphate <0.02% DHEA sodium sulfate <0.02% 17 alpha-hydroxyepipregnanolone <0.02% 19-hydroxyandrostendione <0.02% 17 alpha-hydroxyprogesterone <0.02% Cortisol <0.02% |
| Target Species | Universal |
| For Use With (Application) | ELISA |
| Quantity | 5 Kit |
| Storage Requirements | Store at 4°C. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
