Learn More
Novus Biologicals™ Non-species specific SARS-CoV-2 Spike ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Gene ID (Entrez) | 43740568 |
| Gene Symbol | S |
| Quantity | 1 Kit |
| Research Discipline | Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein |
| Target | Non-species specific |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.