Learn More
Cincinnati Surgical Non-Sterile Carbon Blades
SDP
Packaged with VCI Paper to protect blade from corrosion
$131.50 - $179.20
Specifications
Blade Material British Carbon Steel
Sterility Non-sterile
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-200-261
SDP
View Documents
Cincinnati Surgical
0222
22
Pack of 100 for $131.50
Only null left
Add to Cart
19-200-256
SDP
View Documents
Cincinnati Surgical
0270
70
Pack of 100 for $179.20
Only null left
Add to Cart
Description
Description
  • Made from high-grade British surgical carbon steel
  • Narrow grinding angle resulting in increased initial keenness
  • Medium surface finish
  • 200% visual inspection
  • Strong foil package
Specifications
Specifications
British Carbon Steel
Non-sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.