Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonacosane, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7188029
Click to view available options
Quantity:
250 mg
1 g

Nonacosane,an aliphatic hydrocarbon, is important constituent of the leaf oil of Syzygium cumini tree, possessing anti-bacterial, anti-inflammatory and anti-ulcergenic properties.

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 630-03-5
Physical Form Neat
Linear Formula CH3(CH2)27 CH3
MDL Number MFCD00009356
Melting Point 63°C to 66°C; 63°C to 66°C
Boiling Point 286°C (15 mmHg)
Type Analytical Standard
Quantity 250 mg
Formula Weight 408.79
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.