Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonadecylbenzene 98.0+%, TCI America™
SDP

Catalog No. N034925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N034925G 25 g
N03495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N034925G Supplier TCI America Supplier No. N034925G

Chemical Identifiers

CAS 29136-19-4
Molecular Formula C25H44
Molecular Weight (g/mol) 344.627
MDL Number MFCD00026692
InChI Key SHWJJBRTHGGZBE-UHFFFAOYSA-N
Synonym 1-Phenylnonadecane
PubChem CID 94400
IUPAC Name nonadecylbenzene
SMILES CCCCCCCCCCCCCCCCCCCC1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 344.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Nonadecylbenzene
EINECSNumber (3)-0021
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.