Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonadecylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. N039425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N039425G 25 g
N03945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N039425G Supplier TCI America Supplier No. N039425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22349-03-7
Molecular Formula C25H50
Molecular Weight (g/mol) 350.68
MDL Number MFCD00058954
InChI Key YVTPTBVYGTYTKP-UHFFFAOYSA-N
Synonym 1-Cyclohexylnonadecane
PubChem CID 89671
IUPAC Name nonadecylcyclohexane
SMILES CCCCCCCCCCCCCCCCCCCC1CCCCC1

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 420°C
Quantity 25 g
Formula Weight 350.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Nonadecylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.