Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonane, 99%, Reagent Grade, Solstice

Catalog No. 60048100 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-100 100 mL Glass bottle
60-048-102 500 mL Glass bottle
60-048-101 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 60-048-100 Supplier Solstice Supplier No. N29406100ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-84-2
Molecular Formula C9H20
Molecular Weight (g/mol) 128.259
MDL Number MFCD00009574
InChI Key BKIMMITUMNQMOS-UHFFFAOYSA-N
Synonym n-nonane, shellsol 140, nonyl hydride, nonan, heptane, ethyl, unii-t9w3vh6g10, ccris 6081, hsdb 107, dsstox_cid_5796, nonane, analytical standard
PubChem CID 8141
ChEBI CHEBI:32892
IUPAC Name nonane
SMILES CCCCCCCCC

Specifications

Melting Point -53°C
Color Colorless
Density 0.718g/mL at 25°C
Boiling Point 151°C (1013 hPa)
Flash Point 31°C (87.8°F)
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3(CH2)7CH3
Quantity 100 mL
UN Number UN1920
Beilstein 1696917
Formula Weight 128.26g/mol
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material Nonane
Show More Show Less
Hazard Statement P210-P273-P301 + P310-P304 + P340 + P312-P331-P391
DOTInformation 3/PGIII
RTECSNumber 203-913-4
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.