Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nonanoyl Chloride 95.0+%, TCI America™

Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 215°C |
| Quantity | 100 g |
| UN Number | 3265 |
| Formula Weight | 176.68 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | Nonanoyl Chloride |
Safety and Handling
| EINECSNumber | (2)-0632 |
| TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction