Learn More
NONO Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI2C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504830

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an RNA-binding protein which plays various roles in the nucleus, including transcriptional regulation and RNA splicing. A rearrangement between this gene and the transcription factor E3 gene has been observed in papillary renal cell carcinoma. Alternatively spliced transcript variants have been described. Pseudogenes exist on Chromosomes 2 and 16.Specifications
NONO | |
Monoclonal | |
Unconjugated | |
NONO | |
MRXS34, NMT55, NRB54, P54, P54NRB, PPP1R114 | |
Mouse | |
Affinity Chromatography | |
RUO | |
317259, 4841, 53610, 612773 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15233, Q5FVM4, Q99K48 | |
Nono | |
Human recombit protein fragment corresponding to amino acids 184-385 of human NONO produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.