Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NONO Mouse anti-Human, Mouse, Rat, Clone: 2A2B10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | NONO |
|---|---|
| Clone | 2A2B10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6791
![]()
|
Proteintech Group Inc
663611IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes an RNA-binding protein which plays various roles in the nucleus, including transcriptional regulation and RNA splicing. A rearrangement between this gene and the transcription factor E3 gene has been observed in papillary renal cell carcinoma. Alternatively spliced transcript variants have been described. Pseudogenes exist on Chromosomes 2 and 16.Specifications
| NONO | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 55 kDa nuclear protein, NONO, NonO protein, NRB54, P54, p54(nrb), P54NRB | |
| Nono | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2A2B10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q15233, Q5FVM4, Q99K48 | |
| 317259, 4841, 53610 | |
| NONO Fusion Protein Ag17609 | |
| Primary | |
| -20°C | |
| NONO |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title