Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NonO Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993265
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3265 10 μg
50-199-3264 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3265 Supplier Active Motif Supplier No. 61248
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes an RNA-binding protein which plays various roles in the nucleus, including transcriptional regulation and RNA splicing. A rearrangement between this gene and the transcription factor E3 gene has been observed in papillary renal cell carcinoma. Alternatively spliced transcript variants have been described. Pseudogenes exist on Chromosomes 2 and 16.

Specifications

Antigen NonO
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene Nono
Gene Accession No. Q15233
Gene Alias 54 kDa nuclear RNA- and DNA-binding protein; 55 kDa nuclear protein; AA407051; AV149256; DNA-binding p52/p100 complex, 52 kDa subunit; HGNC:7871; LOC100346936; LOC100348685; MRXS34; NMT-1; NMT55; nonA; Nono; NonO protein; non-POU domain containing, octamer-binding; non-POU domain containing, octamer-binding-like; non-POU domain-containing octamer (ATGCAAAT) binding protein; Non-POU domain-containing octamer-binding protein; non-POU-domain-containing; non-POU-domain-containing, octamer binding protein; non-POU-domain-containing, octamer-binding protein; NRB54; P54; p54(nrb); p54nrb; PPP1R114; protein phosphatase 1, regulatory subunit 114; splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)-like
Gene Symbols Nono
Host Species Rabbit
Immunogen This NonO antibody was raised against a peptide corresponding to the C-terminus of human NonO.
Purification Method Protein A
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4841
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.