Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Nonradioactive Phosphatase Assay Systems

Catalog No. PRV2460 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
Ser or Thr system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRV2460 Ser or Thr system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV2460 Supplier Promega Supplier No. V2460
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Non-Radioactive Phosphatase Assay Systems provide a fast, convenient and flexible alternative for measuring protein phosphatase activity.

  • Absorbance-Based Detection of Phosphatase Activity
  • Choose from serine/threonine or tyrosine phosphatase assays
  • Use with purified samples, crude cell or tissue extracts
  • Convenient alternative to radioactive phosphatase assays

Specifications

For Use With (Application) Can be used with partially fractionated and purified samples as well as in crude cell or tissue extracts
Content And Storage 2 to 10°C
Product Type Ser or Thr system
Quantity 96 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.