Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNase-free Microfuge Tubes

Catalog No. 50591363
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Surface Type:
Graduated
Graduated, low-binding
Quantity:
1000 Tubes/Pk.
250 Tubes/Pk.
500 Tubes/Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Surface Type Quantity
50-591-363 1.5 mL Graduated, low-binding 250 Tubes/Pk.
AM12350 0.5 mL Graduated, low-binding 500 Tubes/Pk.
AM12300 0.5 mL Graduated, low-binding 1000 Tubes/Pk.
AM12400 1.5 mL Graduated, low-binding 500 Tubes/Pk.
AM12475 2 mL Graduated, low-binding 250 Tubes/Pk.
AM12425 2 mL Graduated 500 Tubes/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 50-591-363 Supplier Invitrogen™ Supplier No. AM12450
Add to Cart
Edge
Add to Cart

These microcentrifuge tubes are guaranteed RNase- and DNase-free.

Invitrogen microcentrifuge tubes are guaranteed RNase- and DNase-free.

Features include:

  • Marked with graduations for convenience
  • Flat lids for easy labeling
  • Positive seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Capacity (Metric) 1.5 mL
Material Polypropylene
Quantity 250 Tubes/Pk.
Content And Storage Store at room temperature.
Product Line Invitrogen
Shipping Condition Room Temperature
Dnase Free RNase-Free, DNase-Free
Autoclavable Autoclavable
Closure Type Snap Cap
Graduated Yes
Product Type Microfuge Tube (RNase-free)
Surface Type Graduated, low-binding
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.