Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonyl 4-Hydroxybenzoate 98.0+%, TCI America™
SDP

Catalog No. H0217100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0217100G 100 g
H021725G 25 g
H0217500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0217100G Supplier TCI America Supplier No. H0217100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38713-56-3
Molecular Formula C16H24O3
Molecular Weight (g/mol) 264.365
MDL Number MFCD00016483
InChI Key MBNSKHJDYXPONL-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid Nonyl Ester, Nonylparaben
PubChem CID 586708
IUPAC Name nonyl 4-hydroxybenzoate
SMILES CCCCCCCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 44°C
Color White
Quantity 100 g
Formula Weight 264.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nonyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.