Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. N037925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N037925ML 25 mL
N03795ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N037925ML Supplier TCI America Supplier No. N037925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2883-02-5
Molecular Formula C15H30
Molecular Weight (g/mol) 210.41
MDL Number MFCD00058955
InChI Key CLMFECCMAVQYQA-UHFFFAOYSA-N
Synonym 1-Cyclohexylnonane
PubChem CID 17900
IUPAC Name nonylcyclohexane
SMILES CCCCCCCCCC1CCCCC1

Specifications

Color Colorless
Boiling Point 282°C
Quantity 25 mL
Formula Weight 210.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nonylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.