Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOP1 Monoclonal Antibody (28F2)
GREENER_CHOICE

Catalog No. 01670282
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-282 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-282 Supplier Invitrogen™ Supplier No. MA110025
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10025 detects Yeast NOP-1 from S. cerevisiae samples and does not cross-react with fibrillarin in other species. For chemiluminescent Western blot using yeast samples, use a dilution of 1:10,000 for cell lysates and 1:25,000 for nuclear fractions. For non-ECL detection methods, use a dilution of 1:1000-1:5000.

Nop1p is the yeast homologue of human fibrillarin, a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 28F2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 5mM sodium azide
Gene NOP1
Gene Accession No. P15646
Gene Alias D2870; Histone-glutamine methyltransferase; LOT3; NOP1; Nop1p; Nucleolar protein 1; rRNA 2'-O-methyltransferase fibrillarin; rRNA methyltransferase NOP1; U3 small nucleolar RNA-associated protein NOP1; YDL014W
Gene Symbols NOP1
Host Species Mouse
Immunogen Yeast nuclear preparation.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 851548
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.