Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOP14 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222227JF646
Description
NOP14 Polyclonal antibody specifically detects NOP14 in Human, Mouse samples. It is validated for Western BlotSpecifications
NOP14 | |
Polyclonal | |
Western Blot | |
C4orf9, chromosome 4 open reading frame 9, NOL14nucleolar protein 14 homolog (yeast), NOP14 homolog, NOP14 nucleolar protein homolog (yeast), Nucleolar complex protein 14, nucleolar protein 14, nucleolar protein 14 homolog, probable nucleolar complex protein 14, RES425, RES4-25 | |
The immunogen this antibody was made to, maps to a region between residue 750 and 800 of human Nucleolar Complex Protein 14 using the numbering given in entry NP_003694.1 (GeneID 8602). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
8602 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction