Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOP14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565964 Supplier Invitrogen™ Supplier No. PA565964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQELCQSTLTE MESQKQLCRP LTCEKSKPVP LKLFTPRLVK VLEFGRKQGS SKEEQERKRL IHKHKREFKG AVREIRKDNQ FLARMQLSEI MER Highest antigen sequence identity to the following orthologs - mouse 91%, rat 90%.

Involved in nucleolar processing of pre-18S ribosomal RNA. Has a role in the nuclear export of 40S pre-ribosomal subunit to the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP14
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOP14
Gene Accession No. P78316
Gene Alias 2610033H07Rik; C4orf9; NOL14; NOP14; NOP14 nucleolar protein; NOP14 nucleolar protein homolog; nucleolar complex protein 14; Nucleolar protein 14; probable nucleolar complex protein 14; RES425; RES4-25; RGD1305605; UTP2
Gene Symbols NOP14
Host Species Rabbit
Immunogen Recombinant Human NOP14. Recombinant protein control fragment (Product #RP-106654).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8602
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.