Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOP14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103985 Supplier Invitrogen™ Supplier No. PA5103985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NOL14.

Involved in nucleolar processing of pre-18S ribosomal RNA. Has a role in the nuclear export of 40S pre-ribosomal subunit to the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NOP14
Gene Accession No. P78316, Q8R3N1
Gene Alias 2610033H07Rik; C4orf9; NOL14; NOP14; NOP14 nucleolar protein; NOP14 nucleolar protein homolog; nucleolar complex protein 14; Nucleolar protein 14; probable nucleolar complex protein 14; RES425; RES4-25; RGD1305605; UTP2
Gene Symbols NOP14
Host Species Rabbit
Immunogen A synthesized peptide derived from human NOP14(Accession P78316), corresponding to amino acid residues M338-S388.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289724, 75416, 8602
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.