Learn More
Invitrogen™ NOP14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587842
Description
Immunogen sequence: MAKAKKVGAR RKASGAPAGA RGGPAKANSN PFEVKVNRQK FQILGRKTRH DVGLPGVSRA RALRKRTQTL LKEYKERDKS NVFRDKRFGE YNSNMSPEEK MMKRFALEQQ RHHEKKSIYN LNEDEELTHY GQSLADIEKH NDIVDSDSDA EDRGTLSAEL TAAHFGGGGG LLHKKTQQEG EEREKPKSRK ELIEELIAKS KQEKRERQAQ REDALELTEK LDQDWKEIQT LLSHKTPKSE; Positive Samples: HeLa, U-87MG, HT-29, Jurkat, MCF7; Cellular Location: Nucleus, nucleolus.
Involved in nucleolar processing of pre-18S ribosomal RNA. Has a role in the nuclear export of 40S pre-ribosomal subunit to the cytoplasm.
Specifications
NOP14 | |
Polyclonal | |
Unconjugated | |
NOP14 | |
2610033H07Rik; C4orf9; NOL14; NOP14; NOP14 nucleolar protein; NOP14 nucleolar protein homolog; nucleolar complex protein 14; Nucleolar protein 14; probable nucleolar complex protein 14; RES425; RES4-25; RGD1305605; UTP2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8602 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.12 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78316 | |
NOP14 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human NOP14 (NP_0012789081). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.