Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOP16/HSPC111 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569603
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9603 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9603 Supplier Bethyl Laboratories, Inc Supplier No. A305124AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

NOP16 is transcriptionally regulated by c-Myc (MYC; MIM 190080), upregulated in breast cancer, and overexpression is associated with poor patient survival (Butt et al., 2008).

Specifications

Antigen NOP16/HSPC111
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NOP16
Gene Accession No. Q9Y3C1
Gene Alias HBV pre-S2 trans-regulated protein 3, HSPC111, HSPC185, hypothetical protein HSPC111, LOC51491, NOP16 nucleolar protein homolog, NOP16 nucleolar protein homolog (yeast), nucleolar protein 16 homolog, nucleolar protein 16 homolog (yeast)
Gene Symbols NOP16
Host Species Rabbit
Immunogen Between 25 and 75
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51491
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.