Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOP16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557693 Supplier Invitrogen™ Supplier No. PA557693
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRKRKVKAME VDIEERPKEL VRKPYVLNDL EAEASLPEKK GNTLSRDLID YVRYMVENHG EDYKAMAR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

NOP16 is transcriptionally regulated by c-Myc (MYC), upregulated in breast cancer, and overexpression is associated with poor patient survival (Butt et al., 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP16
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nop16
Gene Accession No. Q1RP77, Q9CPT5, Q9Y3C1
Gene Alias AA409471; CGI-117; D13Wsu177e; HBV pre-S2 trans-regulated protein 3; HSPC111; HSPC185; hypothetical protein HSPC111; Nop16; NOP16 nucleolar protein; NOP16 nucleolar protein homolog; nucleolar protein 16; nucleolar protein 16 homolog; nucleolar protein 16-like; Protein ZH1; RGD1305727
Gene Symbols Nop16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOP16. Recombinant protein control fragment (Product #RP-96803).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28126, 306768, 51491
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.