Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534712 Supplier Invitrogen™ Supplier No. PA534712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji, K562. Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (84%), Pig (97%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NOP2 is a member of the unc-84 homolog family and encodes a nuclear nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Nop2
Gene Accession No. P46087
Gene Alias 120kDa; 25S rRNA (cytosine(2870)-C(5))-methyltransferase; A530002O17; cb164; im:6893299; MGC117384; MGC149287; MGC149288; N2428; NOL1; NOL1/NOP2/Sun domain family, member 1; NOP120; NOP2; NOP2 nucleolar protein; NOP2 nucleolar protein homolog; NOP2 nucleolar protein homolog (yeast); nop2p; NSUN1; Nucleolar protein 1; nucleolar protein 1, 120 kDa; nucleolar protein 1, 120kDa; Nucleolar protein 2; Nucleolar protein 2 homolog; p120; probable 28S rRNA (cytosine(4447)-C(5))-methyltransferase; probable 28S rRNA (cytosine-C(5))-methyltransferase; proliferating-cell nucleolar antigen p120; Proliferation-associated nucleolar protein p120; putative ribosomal RNA methyltransferase NOP2; rRNA (cytosine-C5-)-methyltransferase NOP2; sb:cb164; si:dkey-91i17.1; Unknown (protein for IMAGE:6909257); YNA1; YNL061W; YNL2428W
Gene Symbols Nop2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NSUN1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4839
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.