Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114297
Description
Immunogen sequence: PEQPFEKAAF QKQNDTPKGP QPPTVSPIRS SRPPPAKRKK SQSRGNSQLL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
NOP2 is a member of the unc-84 homolog family and encodes a nuclear nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Alternatively spliced transcript variants have been described.
Specifications
NOP2 | |
Polyclonal | |
Unconjugated | |
Nop2 | |
120kDa; 25S rRNA (cytosine(2870)-C(5))-methyltransferase; A530002O17; cb164; im:6893299; MGC117384; MGC149287; MGC149288; N2428; NOL1; NOL1/NOP2/Sun domain family, member 1; NOP120; NOP2; NOP2 nucleolar protein; NOP2 nucleolar protein homolog; NOP2 nucleolar protein homolog (yeast); nop2p; NSUN1; Nucleolar protein 1; nucleolar protein 1, 120 kDa; nucleolar protein 1, 120kDa; Nucleolar protein 2; Nucleolar protein 2 homolog; p120; probable 28S rRNA (cytosine(4447)-C(5))-methyltransferase; probable 28S rRNA (cytosine-C(5))-methyltransferase; proliferating-cell nucleolar antigen p120; Proliferation-associated nucleolar protein p120; putative ribosomal RNA methyltransferase NOP2; rRNA (cytosine-C5-)-methyltransferase NOP2; sb:cb164; si:dkey-91i17.1; Unknown (protein for IMAGE:6909257); YNA1; YNL061W; YNL2428W | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4839 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P46087 | |
Nop2 | |
Synthetic peptide directed towards the C terminal region of human NOP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction