Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726316 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6316 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6316 Supplier Proteintech Group Inc Supplier No. 104481AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in the regulation of the cell cycle and the increased nucleolar activity that is associated with the cell proliferation. May act as ribosomal RNA methyltransferase.

Specifications

Antigen NOP2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NOP2
Gene Accession No. P46087, Q922K7
Gene Alias NOL1, NOP120, NOP2, NSUN1, Nucleolar protein 1, Nucleolar protein 2 homolog, p120
Gene Symbols Nop2
Host Species Rabbit
Immunogen NOP2 Fusion Protein Ag0498
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110109, 314969, 4839
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.