Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nop25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551958 Supplier Invitrogen™ Supplier No. PA551958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HQEYLKMLAE REEALEEADE LDRLVTAKTE SVQYDHPNHT VTVTTISDLD LSGARLLGLT PPEGGAGDRS EEEASSTEKP TKALPRKSRD PLLSQRISSL TASLHAHSRK KVKRKHPRRA QDSKKPPRAP RTSKAQRRRL TGKAR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

Nop25 is a nucleolar protein predominantly involved in the architecture and function of the nucleolus, which is the cellular hub for ribosomal RNA (rRNA) synthesis and ribosome assembly. It is identified as a putative RNA-binding protein and plays a crucial role in rRNA transcription. Nop25 has been characterized through molecular cloning, showing its ability to bind single-stranded nucleic acids directly. Experimental studies, such as those involving mouse embryogenesis, have utilized techniques like RT-PCR to analyze Nop25's gene expression across developmental stages. Moreover, research on the protein's structure has unveiled its nuclear and nucleolar targeting sequences, vital for its localization to the nucleolus. Treatment of cells with RNase A has illustrated the dissociation of Nop25 from the nucleolus, further highlighting its RNA-binding capabilities. This protein's functional importance is underscored by its role in maintaining nucleolar architecture, as demonstrated by experiments where overexpression of Nop25's N-terminal amino acids disrupted nucleolar integrity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nop25
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOL12
Gene Accession No. Q9UGY1
Gene Alias 25 kDa nucleolar protein; C78541; dJ37E16.7; Nol12; Nop25; Nucleolar protein 12; nucleolar protein of 25 kDa; nucleolar protein, 25 kDa; RGD1305327
Gene Symbols NOL12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Nop25. Recombinant protein control fragment (Product #RP-91493).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79159
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.