Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOP56 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578329
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578329 Supplier Invitrogen™ Supplier No. PA578329
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Chimpanzee (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ErbB 4 (Her-4, c-erbB-4, v-erbB-4) is a member of the class I receptor kinase family. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. ErbB 4 is activated by neuregulins and NTAK (neural and thymus derived activator for ErbB kinases). ErbB 4 is a receptor for heregulin and is capable of mediating HGL-stimulated tyrosine phosphorylation of itself. Like other protein kinases, ErbB 4 mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. ErbB 4 is most predominantly expressed in several breast carcinoma cell lines, and in normal skeletal muscle, heart, pituitary, brain, and cerebellum. Breast tumor cell lines T47-D, MDA-MB-453, BT-474 and H3396 are found to have the highest levels of mRNA, and intermediate levels are seen in MCF-7, MDAMB-330 and MDA-MB-361. Expression of ErbB 4 is low or absent in some breast tumor cell lines such as MDA-MB-231, MDA-MB-157, MDA-MB-468, and SKBR-3. ErbB 4 levels have been found to be elevated in certain human tumor cell lines suggesting that it may play a role in some malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP56
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Nop56
Gene Accession No. O00567
Gene Alias 2310044F10Rik; 56kDa with KKE/D repeat; hypothetical protein 1195; NOL5A; Nop56; NOP56 ribonucleoprotein; NOP56 ribonucleoprotein homolog; nucleolar protein 56; Nucleolar protein 5A; nucleolar protein 5A (56kDa with KKE/D repeat); SCA36; similar to nucleolar protein 5A (56kDa with KKE/D repeat)
Gene Symbols Nop56
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus of human NOL5A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10528
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.