Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOP58 Antibody (34B12), Novus Biologicals™
SDP

Catalog No. NB018364 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018364 0.125 mL
NBP105225 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018364 Supplier Novus Biologicals Supplier No. NBP1052250.125ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NOP58 Monoclonal specifically detects NOP58 in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen NOP58
Applications Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 34B12
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:1000-1:10000, Immunoprecipitation 2-4 ul
Formulation Tissue culture supernatant with 10mM Sodium Azide
Gene Alias HSPC120, NOL5, NOP5/NOP58, NOP58 ribonucleoprotein homolog (yeast), NOP5nucleolar protein 58, Nucleolar protein 5, nucleolar protein NOP5/NOP58
Gene Symbols NOP58
Host Species Mouse
Immunogen A yeast nucleolar preparation
Molecular Weight of Antigen 57 kDa
Purification Method Tissue culture supernatant
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 51602
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.