Learn More
Description
Specifications
Specifications
| Antigen | NOP58 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 34B12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:1000-1:10000, Immunoprecipitation 2-4 ul |
| Formulation | Tissue culture supernatant with 10mM Sodium Azide |
| Gene Alias | HSPC120, NOL5, NOP5/NOP58, NOP58 ribonucleoprotein homolog (yeast), NOP5nucleolar protein 58, Nucleolar protein 5, nucleolar protein NOP5/NOP58 |
| Gene Symbols | NOP58 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
