Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOP58 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100645 Supplier Invitrogen™ Supplier No. PA5100645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NOP58.

Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOP58
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NOP58
Gene Accession No. Q6DFW4, Q9QZ86, Q9Y2X3
Gene Alias HSPC120; MSSP; Nap65; Nol5; NOP5; NOP5/NOP58; Nop58; NOP58 ribonucleoprotein; NOP58 ribonucleoprotein homolog; Nopp140 associated protein; nopp140-associated protein of 65 kDa; Nucleolar protein 5; nucleolar protein 58; nucleolar protein NOP5/NOP58; nucleolar protein NOP58; O6108; RNA-processing protein NOP58; SIK; SIK similar protein; SIK-similar protein; YOR310C
Gene Symbols NOP58
Host Species Rabbit
Immunogen A synthesized peptide derived from human NOP58(Accession Q9Y2X3), corresponding to amino acid residues T409-A459.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51602, 55989, 60373
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.