Learn More
NOP58 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302718AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.Specifications
NOP58 | |
Polyclonal | |
Unconjugated | |
NOP58 | |
HSPC120, NOP5, NOP5/NOP58, NOP58 ribonucleoprotein homolog, NOP58 ribonucleoprotein homolog (yeast), nucleolar protein 5, nucleolar protein NOP5/NOP58 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51602 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9Y2X3 | |
NOP58 | |
Between 350 and 400 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.