Learn More
NOP58 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302719A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.Specifications
NOP58 | |
Polyclonal | |
Unconjugated | |
NOP58 | |
HSPC120, NOP5, NOP5/NOP58, NOP58 ribonucleoprotein homolog, NOP58 ribonucleoprotein homolog (yeast), nucleolar protein 5, nucleolar protein NOP5/NOP58 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51602 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y2X3 | |
NOP58 | |
Between 479 and 529 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.