Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nope/IGDCC4 Antibody (2516A) [DyLight 650], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10345W
Description
Nope/IGDCC4 Monoclonal antibody specifically detects Nope/IGDCC4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Nope/IGDCC4 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
57722 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
2516A | |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
DDM36, hDDM36, immunoglobulin superfamily DCC subclass member 4, immunoglobulin superfamily, DCC subclass, member 4, KIAA1628, likely ortholog of mouse neighbor of Punc E11, LOC57722, Neighbor of punc e11, NOPEFLJ42051, Protein DDM36 | |
Chinese hamster ovary cell line CHO-derived recombinant human Nope/IGDCC4, Met1-Thr957, Accession # Q8TDY8 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction