Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nope Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552580
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552580 Supplier Thermo Scientific Supplier No. PA552580
Add to Cart
Add to Cart

Nope Polyclonal Antibody

Nope Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nope
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IGDCC4
Gene Accession No. Q8TDY8
Gene Alias amplimer WI-16786, mouse homolog; amplimer WI-18508, mouse homolog; DDM36; hDDM36; Igdcc4; Immunoglobulin superfamily DCC subclass member 4; immunoglobulin superfamily, DCC subclass, member 4; Kiaa1628; likely ortholog of mouse neighbor of Punc E11; LOC57722; neighbor of Punc E11; neighbor of Punc e11 protein; Nope; NOPEFLJ42051; Protein DDM36
Gene Symbols IGDCC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Nope. Recombinant protein control fragment (Product #RP-90078).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57722
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.