Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOR-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568825 Supplier Invitrogen™ Supplier No. PA568825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Guinea Pig: 93%; Horse: 93%; Human: 93%; Mouse: 100%; Rabbit: 86%; Rat: 100%.

NOR-1, a NR4 nerve growth factor-like receptor, has been shown to affect neuronal development, T-cell apoptosis, and cancer development. NOR-1 binds to NBRE (NGFI-B response element) and regulates the pro-opiomelanocortin (POMC) gene. NOR-1 is induced rapidly but transiently by a variety of stimuli. Downregulation of NOR-1 and other NGFI-B family members is correlated with drug addiction-prone behavior of Lewis rats. Three human protein isoforms have been documented: NOR-1, NOR-1beta, and NOR-1 3' variant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOR-1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NR4A3
Gene Accession No. Q9QZB6
Gene Alias AI573420; CHN; chondrosarcoma, extraskeletal myxoid, fused to EWS; CSMF; MINOR; mitogen-induced nuclear orphan receptor; Neuron-derived orphan receptor 1; neuron-derived orphan receptor 1/2; Nor1; NOR-1; NOR-2; Nr4a3; Nuclear hormone receptor NOR-1; nuclear hormone receptor NOR-1/NOR-2; Nuclear receptor subfamily 4 group A member 3; nuclear receptor subfamily 4, group A, member 3; Orphan nuclear receptor TEC; TEC; TEC receptor; Translocated in extraskeletal chondrosarcoma
Gene Symbols NR4A3
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse NOR-1 (aa 551-600).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18124
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.