Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOR-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA124537
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA124537 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA124537 Supplier Invitrogen™ Supplier No. PA124537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-24537 detects NOR-1 in human samples.

NOR-1, a NR4 nerve growth factor-like receptor, has been shown to affect neuronal development, T-cell apoptosis, and cancer development. NOR-1 binds to NBRE (NGFI-B response element) and regulates the pro-opiomelanocortin (POMC) gene. NOR-1 is induced rapidly but transiently by a variety of stimuli. Downregulation of NOR-1 and other NGFI-B family members is correlated with drug addiction-prone behavior of Lewis rats. Three human protein isoforms have been documented: NOR-1, NOR-1beta, and NOR-1 3' variant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOR-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NR4A3
Gene Accession No. Q92570
Gene Alias AI573420; CHN; chondrosarcoma, extraskeletal myxoid, fused to EWS; CSMF; MINOR; mitogen-induced nuclear orphan receptor; Neuron-derived orphan receptor 1; neuron-derived orphan receptor 1/2; Nor1; NOR-1; NOR-2; Nr4a3; Nuclear hormone receptor NOR-1; nuclear hormone receptor NOR-1/NOR-2; Nuclear receptor subfamily 4 group A member 3; nuclear receptor subfamily 4, group A, member 3; Orphan nuclear receptor TEC; TEC; TEC receptor; Translocated in extraskeletal chondrosarcoma
Gene Symbols NR4A3
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide near C-terminus of human NR4A3.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100684628, 8013
Target Species Human, Canine, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.