Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NORE1 Antibody (OTI1H2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273041
Description
NORE1 Monoclonal specifically detects NORE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
NORE1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500, Flow Cytometry 1:100, CyTOF-ready | |
Maxp1, MGC10823, MGC17344, New ras effector 1, NORE1A, NORE1B, NORE1RASSF3, Rap1-binding protein, RAPLnovel Ras effector 1, Ras association (RalGDS/AF-6) domain family 5, Ras association (RalGDS/AF-6) domain family member 5, ras association domain-containing protein 5, Ras effector-like protein, regulator for cell adhesion and polarization enriched in lymphoid tissue, Regulator for cell adhesion and polarization enriched in lymphoid tissues, tumor suppressor RASSF3 | |
Mouse | |
30.2 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, CyTOF | |
OTI1H2 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
RASSF5 | |
Full length human recombinant protein of human RASSF5 (NP_872606) produced in HEK293T cell. | |
Affinity Purified | |
Cancer, Signal Transduction, Tumor Suppressors | |
83593 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction